Learn More
Aat Bioquest 8-Hydroxypyrene-1,3,6-trisulfonic acid, trisodium salt [HPTS] [CAS 6358-69-6]
SDP
Supplier:  Aat Bioquest 85
Encompass_Preferred
8-Hydroxypyrene-1,3,6-trisulfonic acid, trisodium salt [HPTS] is the cell-impermeable fluorescent pH indicator used for slightly acidic pH range (pKa = 6.4).
Catalog No. 50-297-9310