Learn More
Sklar Instruments Forceps, Adlerkreutz Tissue Forceps, Sklar 8 in., 2x3 Teeth, Straight, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 191916
Encompass_Preferred
Forceps, Adlerkreutz Tissue Forceps, Sklar, 8 in., 2x3 Teeth, Straight, Stainless Steel, EA. Sklar Alderkreutz Tissue Forceps are thumb forceps used for gripping and handling tissue. They are available in two lengths, 6 inches or 8 inches, and featrue 2X3 or 4X5 teeth in addition to partially, horizontally serrated tips. This product is straight with 2X3 teeth, and has an overall length of 8 inches.
Catalog No. 50-109-8016