Learn More
Chemglass Life Sciences Stirrer, High Volume, Digital, Magnetic, 8L, 50-60hz, 100-240V
SDP
Supplier:  Chemglass Life Sciences CLS410808
Encompass_Preferred
Stirrer, High Volume, Digital, Magnetic, 8L, 50-60hz, 100-240V. High Volume magnetic stirrer has a bright digital display to accurately set the stirring speed. SpeedServo™ ensures constant speed even when the viscosity changes. Magnetic stirrers have been designed and manufactured in compliance with the following standards CE, CSA and UL. The MSL Series is designed for high volume stirring/mixing of water-like solutions. Main Applications: High volumes preparations, cell cultures and tissue cultures (with spinner flasks), buffer solution preparations.
Catalog No. 50-197-9478