Learn More
Ace Glass, Inc. 8MM STOPCOCK GB 8MM STOPCOCK GB
SDP
Supplier:  Ace Glass, Inc. 813712
Encompass_Preferred
Glass 8mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.
Catalog No. 50-979-694