Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8PPD-Q | 99.9% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY17170210MG

Encompass_Preferred

8PPD-Q is a para-phenylenediamine quinone used as a rubber antioxidant with pronounced antioxidant and anti-ozone activity. It is used in environmental pollution and toxicity research because it is toxic to aquatic species, particularly salmonids.

  • High purity: 99.88% as reported by the manufacturer.
  • Solid form suitable for laboratory handling and storage.
  • Molecular formula C20H26N2O2 and molecular weight 326.43 g/mol.
  • Available in small pack sizes for analytical work, including 10 mg.
  • Store powder at -20 °C (3 years) or 4 °C (2 years); in solvent store at -80 °C (6 months) or -20 °C (1 month).
  • Hazardous to aquatic life; suitable for environmental toxicity studies and analytical investigations.

Catalog No. 50-002-95391


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.