Learn More
Medchemexpress LLC (8R,9S)-5-fluoro-8-(4-fluorophenyl)-9-(1-methyl-1H-1,2,4-triazol-5-yl)-2,7,8,9-tetrahydro | 1207456-00-5 | 98.3% | 380.35 | 200 MG
Supplier: Medchemexpress LLC HY16106A200MG
This product is an enantiomer of Talazoparib, functioning as a PARP1 inhibitor with an IC50 of 144 nM. It is intended for research use only.
- Inhibits PARP1.
- Shows cellular PARylation inhibition.
- Demonstrates cytotoxicity against BRCA2-deficient cells.
- Potentiates temozolomide-induced cytotoxicity.