Learn More
Medchemexpress LLC 9,10-Anthracenedione, 1,4-bis(butylamino)- | 17354-14-2 | 99.8% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0516
Encompass_Preferred
Solvent Blue 35, also known as Sudan Blue II, is an anthraquinone-based blue solvent dye. It is primarily used for marking specific diesel fuels and for differentiating agricultural, fishery, and automotive fuels.
  • Blue solvent dye
  • Used for marking specific diesel fuels
  • Differentiates agricultural, fishery, and automotive fuels
Catalog No. 50-004-14290