Learn More
AdipoGen 9,10-Anthracenediyl-bis(methylene)dimalonic acid, 307554-62-7, MFCD03095464, 50 mg
SDP
Supplier:  AdipoGen CDXA0183M050
Encompass_Preferred
MF: C22H18O8, MW: 410.37, ≥90%, Appearance: Light brown powder. Reagent for the assay of O2 with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid.
Catalog No. 50-596-133