Learn More
Medchemexpress LLC 9,10-phenanthrenedione | 84-11-7 | MFCD00001163 | 100.0% | 208.21 g/mol | C14H8O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW014507
Encompass_Preferred
9,10-Phenanthrenequinone (9,10-phenanthrenedione) is a high-purity research chemical (C14H8O2, MW 208.21 g/mol) used in biochemical and cellular studies as a claudin-5 inhibitor. It is supplied as a yellow to brown solid and is offered in multiple pack sizes, with Certificate of Analysis and Safety Data Sheet available for quality and safety verification.
  • High purity (99.95% by HPLC) suitable for research applications.
  • Known claudin-5 inhibitor used in cellular and biochemical assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Certificate of Analysis and safety data sheet available for each batch.
  • Chemical formula C14H8O2 and molecular weight 208.21 g/mol.
Catalog No. 50-003-03583