Learn More
Cayman Chemical 9 10DIhydroxystearIc AcId 10mg
SDP
Supplier:  Cayman Chemical 2861210 MG
Encompass_Preferred
An oxidation product of oleic acid; can be formed from oleic acid in HepG2 cells; activates PPARα in CV-1 cells at 50-100 µM; decreases blood glucose levels, increases insulin sensitivity, and decreases body weight in high-fat diet-fed KKAy diabetic mice at 4% in the diet
Catalog No. 50-293-7754