Learn More
Medchemexpress LLC 9,12-octadecadienamide, N-(1-phenylethyl)-, (9Z,12Z)- | 14417-88-0 | 99.9% | 383.61 g·mol⁻¹ | C26H41NO | 50 MG

Supplier: Medchemexpress LLC HY10172250MG
Melinamide is an amide derivative of an unsaturated long-chain fatty acid that inhibits cholesterol absorption (reported IC50 20.9 μM). Supplied as an oil or DMSO solution for research use, it is used in biochemical studies of cholesterol uptake and acyltransferase-related pathways.
- Molecular formula C26H41NO
- Molecular weight 383.61 g·mol⁻¹
- Reported activity: IC50 20.9 μM for cholesterol absorption inhibition
- Purity 99.9%
- Available as oil and 10 mM DMSO solutions in multiple package sizes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.