Learn More
Medchemexpress LLC 9-(2-Carboxy-2-cyanovinyl)julolidine | 142978-18-5 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD00545MG
Encompass_Preferred
9-(2-Carboxy-2-cyanovinyl)julolidine is a fluorescent molecular rotor. It can be used to bind to IgG and Fab, and for preparing antibodies.
  • Fluorescent molecular rotor
  • Can be used to bind to IgG and Fab
  • Used for preparing antibodies
Catalog No. 50-004-10839