Learn More
Medchemexpress LLC 9-(2-deoxy-beta-D-ribofuranosyl)-6-methylpurine | 16006-64-7 | MFCD29059892 | 95.0% | 250.25 g·mol⁻¹ | C11H14N4O3 | 25 MG

Supplier: Medchemexpress LLC HYW39443225MG
9-(2-Deoxy-beta-D-ribofuranosyl)-6-methylpurine is a purine nucleoside analogue used in cancer research. It exhibits antitumor activity through inhibition of DNA synthesis and induction of apoptosis, making it useful for studies of lymphoid malignancies, nucleoside metabolism, and mechanism-of-action experiments.
- Purity: 95.0%.
- Cas number: 16006-64-7.
- Molecular formula: C11H14N4O3.
- Molecular weight: 250.25 g·mol⁻¹.
- Appearance: Off-white to light yellow solid.
- Solubility: Soluble in DMSO (~125 mg/mL).
- Storage: Powder -20°C for long term; store solutions at -80°C for short term.
- Applications: Suitable for antitumor activity assays and nucleoside metabolic studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.