Learn More
Medchemexpress LLC 9-(2-fluorobenzyl)-6-(methylamino)-9H-purine | 101155-02-6 | 95.0% | 257.27 g·mol⁻¹ | C13H12FN5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10011825MG
Encompass_Preferred
BW-A 78U is a small-molecule phosphodiesterase 4 (PDE4) inhibitor with reported in vitro potency (IC50 ≈ 3 μM). It is supplied for research use and is available as a powder and as a 10 mM solution in DMSO. The compound is characterized by formula C13H12FN5, molecular weight 257.27 g·mol⁻¹, and CAS 101155-02-6.
  • PDE4 inhibition for biochemical and cellular assays.
  • Moderate potency (IC50 ≈ 3 μM) suitable for target validation studies.
  • Available in multiple pack sizes and solution format for assay flexibility.
  • Characterized by CAS number 101155-02-6 and molecular formula C13H12FN5.
  • Typical purity around 95% as reported in the safety data sheet.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-15719