Learn More
FILTROUS INC 9-425 BLUE BONDED SCREW TOP
Supplier:  FILTROUS INC ALB091414
Encompass_Preferred
9-425 Blue Bonded Screw Top Autosampler Caps w/ PTFE/Silicone - FiltrousScrew closureBondedPTFE/Silicone septa9mm neck widthCompatible with most organic solvents (until pierced)Not recommended for use with chlorosilanesQuantity of 100 caps per unitFormerly BLB-0902
Catalog No. NC2206040