Learn More
Bio Rad Laboratories 9.5X15.2 FILTER PAPER
Supplier:  Bio Rad Laboratories 1704085
Encompass
Blot absorbent filter paper (thick), is precut cotton fiber paper that is used in the assembly of a gel and membrane sandwich for electrophoretic transfer, or blotting, and gel drying applications.
Catalog No. NC9768147