Learn More
Medchemexpress LLC 9-acetyl-3,4-dihydro-2H-carbazol-1-one | 352553-42-5 | 99.8% | 227.26 g/mol | C14H13NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY119053100MG
Encompass_Preferred
MS7972 is a small-molecule inhibitor that blocks the interaction between human p53 and CREB-binding protein and has been shown to almost completely inhibit this bromodomain interaction at 50 μM. The compound is supplied as a solid and as a DMSO solution for research use, and is intended for laboratory applications only.
  • Blocks p53-CREB protein-protein interaction at low micromolar concentration.
  • Demonstrated activity at 50 μM in biochemical assays.
  • High purity suitable for research (approximately 99.8%).
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Multiple packaging sizes to support assay development and follow-up studies.
  • Stable as a powder at -20°C for long-term storage; in solution store at -80°C for extended periods.
  • Chemical formula C14H13NO2 and molecular weight 227.26 g/mol.
Catalog No. 50-002-29055