Learn More
Medchemexpress LLC 9-amino-6-chloro-2-methoxyacridine | 602166 | MFCD07374304 | 99.6% | 258.70 g·mol⁻1 | C14H11ClN2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11815525MG
Encompass_Preferred
9-Amino-6-chloro-2-methoxyacridine is a cell-permeable, pH-sensitive fluorescent probe used in research to monitor vacuolar and intracellular pH changes, proton gradients, and membrane transport activity. It is commonly applied in fluorescence-based assays to study proton transporters and pH-dependent processes.
  • pH-sensitive fluorescent probe for measuring vacuolar and intracellular pH.
  • Cell-permeable and suitable for live-cell fluorescence assays.
  • Intercalates with nucleic acids, enabling DNA-related staining applications.
  • High purity (≈99.6%) for reproducible experimental results.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-26910