Learn More
Medchemexpress LLC 9-Aminoacridine | 90-45-9 | 99.4% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB142210MM/1ML
Encompass_Preferred
9-Aminoacridine is a fluorescent probe and a pH indicator, used for quantitative determination of transmembrane pH gradients (inside acidic). It acts as an antimicrobial agent by interacting with specific bacterial DNA and disrupting the proton motive force in K. pneumoniae. It is also an HIV-1 inhibitor, preventing HIV LTR transcription and viral replication in infected cell lines. This compound can additionally be used as an adjuvant for Rifampin in the treatment of multidrug-resistant K. pneumoniae infections.
  • Fluorescent probe and pH indicator
  • Determines transmembrane pH gradients
  • Antimicrobial agent
  • Disrupts proton motive force in K. pneumoniae
  • Inhibits HIV LTR transcription and viral replication
  • Adjuvant for Rifampin in multidrug-resistant K. pneumoniae infections
Catalog No. 50-004-13428