Learn More
AdipoGen 9-Anthracenecarbaldehyde
SDP
Supplier:  AdipoGen CDXA0180G500
Encompass_Preferred
Chemical. CAS 642-31-9. Formula C15H10O. MW 206.24. Synthetic. Fluorescent reagents probe, stain, label with reactive functional group for chromatography or spectroscopy. Derivatization agent. Used for membrane research, luminescent detection, molecular biology, protein modifications, protein structural analysis and proteomics. Carbohydrate labels for chromatography.
Catalog No. 50-256-1813