Learn More
Medchemexpress LLC 9-Bromoanthracene-d9 | 183486-02-4 | 266.18 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW009455S5G
Encompass_Preferred
9-Bromoanthracene-d9 is a deuterium-labeled version of 9-Bromoanthracene. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer and an internal standard for quantitative analysis. Its appearance is a solid, light yellow to yellow in color.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52094