Learn More
Medchemexpress LLC 9-Bromonon-1-ene | 89359-54-6 | 205.14 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00639750G
Encompass_Preferred
9-Bromonon-1-ene is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is sold for research purposes only and not for patient use.
  • Biochemical reagent for life science research.
  • Pure form storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-45353