Learn More
Medchemexpress LLC 9-chloro-2-ethyl-1-methyl-3-(2,2,2-trifluoroethyl)-6H-pyrrolo[3,2-f]quinolin-7-one | 917891-35-1 | 99.6% | 342.74 g/mol | C16H14ClF3N2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10357610MM 1ML
Encompass_Preferred
LGD-3303 is a non-steroidal selective androgen receptor modulator (SARM) used in research to study androgen receptor pharmacology. Supplied as a 10 mM solution (1 mL) for laboratory use, it is intended for research use only and not for human or clinical applications.
  • Selective androgen receptor activity for target validation and pharmacology studies.
  • Reported purity of 99.6% suitable for analytical and biological assays.
  • Provided as a concentrated solution to simplify dilution and dosing.
  • Compact 1 mL volume appropriate for small-scale experiments and screening.
Catalog No. 50-002-44619