Learn More
TARGETMOL CHEMICALS INC 9-CP-ADE MESYLATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T235885MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. 9-CP-Ade Mesylate (9 CP Ade Mesylate) is a cell-permeable stable and non-competitive inhibitor of adenylate cyclase. purity: 99%
Catalog No. 50-258-0855