Learn More
Medchemexpress LLC 9-Dihydro-13-acetylbaccatin III | 142203-65-4 | 99.6% | C33H42O12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY774345MG
Encompass_Preferred
9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate used for the preparation of paclitaxel analogs. It has shown antiproliferative activity against mouse P388 cells with an IC50 of 20 μg/mL in an MTT assay.
  • Intermediate for the preparation of paclitaxel analogs
  • Exhibits antiproliferative activity against mouse P388 cells
  • Derived from plants, Taxaceae, Taxus chinensis (Pilger) Rehd.
  • Classified as a diterpenoid
Catalog No. 50-004-33392