Learn More
Medchemexpress LLC 9-Dihydro-13-acetylbaccatin III (9-DHAB III) | 142203-65-4 | 99.6% | C33H42O12 | 100 MG

Supplier: Medchemexpress LLC HY77434100MG
9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate for the preparation of paclitaxel analogs. It has demonstrated antiproliferative activity against mouse P388 cells with an IC50 of 20 μg/mL, as determined by an MTT assay.
- Purity of 99.6%
- Molecular weight: 630.68
- Chemical formula: C33H42O12
- Appearance: White to off-white solid
- Classified as a terpenoid and diterpenoid
- Derived from Taxus chinensis plants
- Soluble in DMSO at 20 mg/mL
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.