Learn More
Medchemexpress LLC 9-fluorenol (9-hydroxyfluorene) | 1689-64-1 | MFCD00001135 | 99.9% | 182.22 g/mol | C13H10O | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01638810G
Encompass_Preferred
9-Fluorenol (9-hydroxyfluorene) is a research-grade small molecule (CAS 1689-64-1) used as a dopamine transporter (DAT) inhibitor and as an analytical standard or metabolite reference. It is supplied as a solid and as a DMSO solution for analytical and in vivo research applications. For research use only; not for diagnostic or therapeutic purposes.
  • High purity (99.87%).
  • Molecular formula C13H10O; molecular weight 182.22 g/mol.
  • Available as solid in multiple gram sizes and as 10 mM solution in DMSO.
  • Suitable as an analytical standard and metabolite reference in pharmacology studies.
  • Provided for research use only.
Catalog No. 50-003-02730