Learn More
Medchemexpress LLC 9-Fluorenylmethanol | 24324-17-2 | 98.8% | 196.25 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW009049100G
Encompass_Preferred
9-Fluorenylmethanol is an N-protecting reagent that can be used in peptide synthesis. It is intended for research use only.
  • Can be used in peptide synthesis.
  • Appearance is solid.
  • Color ranges from off-white to light yellow.
Catalog No. 50-003-66894