Learn More
Medchemexpress LLC 9-fluorenylmethyl carbamate | 84418-43-9 | MFCD00237376 | 99.8% | 239.27 g/mol | C15H13NO2 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW015987500G
Encompass_Preferred
9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected amide used in organic synthesis and as a photobase initiator for preparing organosilane-based proton exchange membranes. Supplied for research use, it has formula C15H13NO2 and a molecular weight of 239.27 g/mol.
  • Provides Fmoc-protected primary amine for peptide and small-molecule synthesis.
  • Functions as a photobase initiator in polymer and membrane research.
  • High purity suitable for research and analytical applications.
  • Solid, stable material with well-defined chemical identity.
Catalog No. 50-002-99673