Learn More
Medchemexpress LLC 9-fluorenylmethyl carbazate | 35661-51-9 | MFCD00239419 | 99.0% | 254.28 | C15H14N2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01787310MM/1ML
Encompass_Preferred
9-Fluorenylmethyl carbazate is a fluorescent labeling reagent used to derivatize glycans and other analytes for fluorimetric detection. It is supplied in solid form and is also available as a 10 mM solution in DMSO, making it suitable for HPLC derivatization and fluorescence-based assays. Recommended storage and stability vary by form and are provided by the manufacturer.
  • Used for fluorimetric detection of glycans.
  • Available as powder and as a 10 mM solution in DMSO.
  • Molecular formula C15H14N2O2; molecular weight 254.28.
  • Suitable for HPLC derivatization and fluorescence assays.
  • Manufacturer provides storage conditions for powder and solutions.
Catalog No. 50-003-01308