Learn More
Medchemexpress LLC HY-N2795 1mg Medchemexpress, 9-Hydroxycalabaxanthone CAS:35349-68-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN27951MG
Encompass_Preferred
Medchemexpress, HY-N2795 1mg 9-Hydroxycalabaxanthone CAS:35349-68-9 9-Hydroxycalabaxanthone (Xanthone I) is a known xanthone isolated from Garcinia mangostana Linn. 9-Hydroxycalabaxanthone has quorum-sensing inhibitory, anti-microbial, and anti-malarial activities (IC50=1.2-1.5 µM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2909