Learn More
Cayman Chemical 9-HydroxyelIptIcInhydroch 10mg
SDP
Supplier:  Cayman Chemical 1792110 MG
Encompass_Preferred
An ellipticine derivative with diverse biological activities; inhibits aroclor-induced activation of aniline hydroxylase, aminopyrine N-demethylase, and 7-ethoxycourmarin O-deethylase in rat liver microsomes (Kis = 3.5, 0.6, and 0.74 μM, respectively); inhibits the growth of L1210 murine leukemia cells (IC50 = 3 nM) in vitro and increases survival in an L1210 mouse leukemia model; inhibits carrageenan-induced edema and UV-induced erythema in guinea pigs
Catalog No. 50-296-6976