Learn More
Medchemexpress LLC 9-ING-41 | 1034895-42-5 | 99.13% | 404.35 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113914100MG
Encompass_Preferred
9-ING-41 (Elraglusib) is a maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor with an IC50 of 0.71 μM. This compound significantly induces cell cycle arrest, autophagy, and apoptosis in cancer cells. It demonstrates anticancer activity and shows potential for enhancing the antitumor effects of chemotherapeutic agents.
  • It is a maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor.
  • It leads to cell cycle arrest, autophagy, and apoptosis in cancer cells.
  • It has anticancer activity.
  • It has the potential for enhancing the antitumor effects of chemotherapeutic agents.
Catalog No. 50-004-38447