Learn More
Medchemexpress LLC 9-Octadecenoic acid (9Z)-, 1,1'-[1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl] ester | 2190-30-9 | 99.8% | 859.39 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11213250MG
Encompass_Preferred
Palmitodiolein (Triglyceride POO) is a triacylglycerol found in vegetable oils. It is for research use only and not sold to patients.
  • Forms an N-nitroso compound after interacting with morpholine in vitro.
Catalog No. 50-004-37212