Learn More
Medchemexpress LLC 9-octadecynoic acid | 506-24-1 | MFCD00014386 | 99.9% | 280.45 g/mol | C18H32O2 | 100 MG

Supplier: Medchemexpress LLC HYW341997100MG
9-Octadecynoic acid is an acetylenic C18 fatty acid supplied for research use. It functions as a DNA-binding agent (Kd = 1.8 mM) and as an agonist of peroxisome proliferator-activated receptor γ (PPARγ), making it useful for studies of lipid signaling, receptor activation, and DNA interactions.
- High chemical purity (99.9%).
- Molecular formula C18H32O2; molecular weight 280.45 g/mol.
- Available as a solid powder suitable for dissolution in organic solvents.
- Storage: powder stable at -20°C; in solvent store at -80°C for longer-term stability.
- Suitable for biochemical assays and cell-based studies involving PPARγ activation or DNA binding.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.