Learn More
Cayman Chemical 9-OxoDE-d3 100ug
SDP
Supplier:  Cayman Chemical 338420100 UG
Encompass_Preferred
An internal standard for the quantification of 9-OxoODE by GC- or LC-MS
Catalog No. 50-294-8312