Learn More
Cayman Chemical 9-OxoDE-d3 50ug
SDP
Supplier:  Cayman Chemical 33842050 UG
Encompass_Preferred
An internal standard for the quantification of 9-OxoODE by GC- or LC-MS
Catalog No. 50-294-9233