Learn More
Medchemexpress LLC 9-(oxolan-2-yl)purin-6-amine | 17318-31-9 | MFCD00210216 | ≥98.0% | 205.22 g/mol | C9H11N5O | 200 MG
SDP
Supplier:  Medchemexpress LLC HY100396200MG
Encompass_Preferred
SQ22536 is a small-molecule adenylate cyclase inhibitor used in research to modulate cAMP signaling. It is provided as a powder and is characterized by CAS 17318-31-9, molecular formula C9H11N5O, and molecular weight 205.22 g/mol. The compound is suitable for laboratory-scale in vitro and in vivo studies with documented solubility and storage recommendations.
  • Potent adenylate cyclase inhibitor for cAMP pathway studies.
  • Suitable for in vitro and in vivo applications with established vehicle formulations.
  • Soluble in DMSO and water at experimentally useful concentrations.
  • Stable as a powder under cold storage with multi-year shelf life.
  • Provided as an analytical standard for research use only.
Catalog No. 50-002-15420