Learn More
Medchemexpress LLC 9-oxooctadecanoic acid | 4114-74-3 | MFCD05864058 | >98.0% | 298.5 g/mol | C18H34O3 | 25 MG

Supplier: Medchemexpress LLC HYW67004425MG
9-Oxooctadecanoic acid is a long-chain oxo fatty acid used as a research reagent in biochemical and metabolic studies. It is the 9-oxo derivative of stearic acid and is used as a reference standard, intermediate, or assay reagent in laboratory work.
- High purity (>98%) suitable for research applications.
- Chemical formula: C18H34O3.
- Molecular weight: 298.5 g/mol.
- CAS number: 4114-74-3.
- Suitable for biochemical assays and metabolic research.
- Supplied in small-scale packages for laboratory use (e.g., 25 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.