Learn More
Cayman Chemical 9 Z 11 E 13 Z-OctadecatrIe 5mg
SDP
Supplier:  Cayman Chemical 260575 MG
Encompass_Preferred
An ω-5 long-chain polyunsaturated fatty acid; has been found as a major component in pomegranate seed oil; binds to the AF2 domain of PPARγ (IC50 = 2.5 µM); increases PPARα and PPARγ activity in 3T3-L1 cells in a reporter assay
Catalog No. 50-293-5893