Learn More
Cayman Chemical 9 Z 11 E 13 Z-OctadecatrIe 5mg
SDP
Supplier:  Cayman Chemical 248075 MG
Encompass_Preferred
A fatty acid methyl ester; isomer of 9(Z),11(E),13(E)-octadecatrienoic acid methyl ester; has been used as a standard for the quantification of 9(Z),11(E),13(Z)-octadecatrienoic acid in wild growing pomegranate seed oil
Catalog No. 50-293-4839