Learn More
Medchemexpress LLC 9(Z),11(E)-conjugated linoleic acid methyl ester | 13058-52-1 | 99.2% | 294.47 g/mol | C19H34O2 | 10MG

Supplier: Medchemexpress LLC HY12437010MG
9(Z),11(E)-Conjugated linoleic acid methyl ester is the methyl ester derivative of a conjugated linoleic acid isomer used as a research reagent and analytical standard for quantification of conjugated fatty acids in lipid studies. It is supplied as a high-purity liquid suitable for chromatography and reference standard applications.
- Used as a reference standard in lipid analysis.
- Molecular formula: C19H34O2.
- Molecular weight: 294.47 g/mol.
- Purity: 99.2%.
- Physical form: liquid.
- Storage: protect from light and store under nitrogen at -20°C (in solvent: -80°C up to 6 months).
- Available in small milligram quantities for analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.