Learn More
Medchemexpress LLC 9(Z)-Octadecenedioic acid | 20701-68-2 | 99.4% | 312.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY166198100MG
Encompass_Preferred
9(Z)-Octadecenedioic acid is a C18 cutin monomer and marine biodegradation rate accelerator for biodegradable resins. It is a structural analog of 9,10,16-trihydroxyhexadecanoic acid. For research use only, it serves as a marine biodegradation rate accelerator for synthesizing biodegradable resins and other materials.
  • C18 cutin monomer
  • Marine biodegradation rate accelerator for biodegradable resins
Catalog No. 50-003-94247