Learn More
Medchemexpress LLC 9(Z)-Octadecenedioic acid | 20701-68-2 | 99.4% | 312.44 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16619850MG
Encompass_Preferred
9(Z)-Octadecenedioic acid is a C18 cutin monomer and a marine biodegradation rate accelerator for biodegradable resins. It is also a structural analog of 9,10,16-trihydroxyhexadecanoic acid.
  • C18 cutin monomer
  • Marine biodegradation rate accelerator for biodegradable resins
  • Structural analog
  • For research use only
Catalog No. 50-003-94646