Learn More
Medchemexpress LLC 9(Z)-Octadecenedioic acid | 20701-68-2 | 99.4% | 312.44 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY166198500MG
Encompass_Preferred
9(Z)-Octadecenedioic acid is a C18 cutin monomer that acts as a marine biodegradation rate accelerator for synthesizing biodegradable resins and other materials. It is also a structural analog of 9,10,16-trihydroxyhexadecanoic acid.
  • C18 cutin monomer
  • Marine biodegradation rate accelerator
  • Structural analog of 9,10,16-trihydroxyhexadecanoic acid
  • Used for synthesizing biodegradable resins
Catalog No. 50-003-94693