Learn More
Cayman Chemical 9 Z-TrIcosen 5g
SDP
Supplier:  Cayman Chemical 132365 G
Encompass_Preferred
An insect pheromone used in pesticides; activates antennal basiconic Or7a receptors in Drosophila; produced by honey bees to communicate information about food sources
Catalog No. 50-295-1912