Learn More
Neuvitro Collagen coated sterile German glass coverslips for cell culture, 22mm dia #1.5 thick 90/pk, ready to use in cell culture.
Supplier:  Neuvitro GG221.5COLLAGEN
Encompass_Preferred
CE certified German glass coverslips with rat tail collagen I coating on both sides for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, coated with pathgen-free rat tail collagen I and sterile. 22mm diameter round, #1.5 thickness, 90 pieces/pack. Ready to use directly in cell culture. Coverslips are sealed as 15 pieces per group for easy storage and enhanced cell culture quality. Shelf life 0.5 year when stored in chest freezer. User manual included.
Catalog No. NC0389465