Learn More
Grainger 90 Sanitary Tee 316 SS 3 in 29 PSI
Supplier:  Grainger 14N983
Encompass_Preferred
The Blucher 90 Sanitary Tee is a stainless steel sanitary tee fitting with a 90 degree angle. It has a 3 inch diameter and is made from 316L stainless steel. The tee has a polished interior finish and is rated for pressures up to 150 psi at temperatures up to 250 degrees Fahrenheit. It meets 3-A sanitary standards and is designed for use in sanitary process piping applications in food, beverage, pharmaceutical, and other industries requiring sanitary process components. The tee provides a 90 degree branch connection from a straight run pipe section.
Catalog No. 50-275-1605