Learn More
eMolecules​ 920-46-7 | Methacryloyl chloride contains ~200ppm monomethyl ether hydroquinone as stabilizer | Oakwood Chemical | MFCD00000716 | 104.530 | C4H5ClO | 97.000 | CC(=C)C(Cl)=O | 1g | 537711988
Supplier:  eMolecules​ 537711988
Encompass_Preferred
Methacryloyl chloride contains ~200ppm monomethyl ether hydroquinone as stabilizer | Oakwood Chemical | 920-46-7 | MFCD00000716 | 104.530 | C4H5ClO | 97.000 | CC(=C)C(Cl)=O | 1g | 537711988
Catalog No. 50-001-58564