Learn More
ATCC 93T449; Liposarcoma; Human (Homo sapiens)
Supplier:  ATCC CRL3043
Encompass
93T449 is a cell line exhibiting fibroblast-like morphology that was isolated in 1993 from the retroperitoneum of a 68-year-old, White female with liposarcoma. The MDM2 amplification present in this cell line can be used as a model for testing new drugs and cancer research. This cell line is one of the rare liposarcoma cell lines available.
Catalog No. 50-238-2389
 Restricted Item